missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL 32A Protein

A cDNA sequence encoding the IL 32A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12398-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990289

Quantity Price
1 171.64 € / 20µg
10 161.27 € / 20µg
Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

9235
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
IL32
Human
His
The IL-32 His Tag protein solution contains 20mM Tris-HCl, pH 8.0, 1mM DTT and 10% glycerol.
IL 32A Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMCF PKVLSDDMKK LKARMHQAIE RFYDKMQNAE SGRGQVMSSL AELEDDFKEGYLETVAAYYE EQHPELTPLL EKERDGLRCR GNRSPVPDVE DPATEEPGES FCDKSYGAPR GDKEELTPQK CSEPQSSK
Videos
SDS
Documents
Certificates