missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL10RA Protein
A cDNA sequence encoding the IL10RA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12401-2ug
Additional Details : Weight : 0.01000kg
Specifications
3587 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
HGTELPSPPS VWFEAEFFHH ILHWTPIPNQ SESTCYEVAL LRYGIESWNS ISNCSQTLSY DLTAVTLDLY HSNGYRARVR AVDGSRHSNW TVTNTRFSVD EVTLTVGSVN LEIHNGFILG KIQLPRPKMA PANDTYESIF SHFREYEIAI RKVPGNFTFT HKKVKHENFS LLTSGEVGEF CVQVKPSVAS RSNKGMWSKE ECISLTRQYF TVTNHHHHHH | |
Greater than 85% as determined by SDS-PAGE. |
IL10RA Protein | |
Research Use Only | |
IL10RA | |
Human | |
His | |
The IL10RA protein solution (0.2 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol. |