missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL17RB Protein
A cDNA sequence encoding the IL17RB was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 55540 |
---|---|
Name | IL17RB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | IL17RB |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15990329
|
enQuireBio™
QP12409-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970329
|
enQuireBio™
QP12409-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15980329
|
enQuireBio™
QP12409-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
55540 | |
Research Use Only | |
IL17RB | |
Human | |
His | |
IL17RB protein solution (0.25 mg/ml) containing phosphate buffered saline (pH 7.4) and 10% glycerol. |
IL17RB Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
REPTVQCGSE TGPSPEWMLQ HDLIPGDLRD LRVEPVTTSV ATGDYSILMN VSWVLRADAS IRLLKATKIC VTGKSNFQSY SCVRCNYTEA FQTQTRPSGG KWTFSYIGFP VELNTVYFIG AHNIPNANMN EDGPSMSVNF TSPGCLDHIM KYKKKCVKAG SLWDPNITAC KKNEETVEVN FTTTPLGNRY MALIQHSTII GFSQVFEPHQ KKQTRASVVI PVTGDSEGAT VQLTPYFPTC GSDCIRHKGT VVLCPQTGVP FPLDNNKSKP GGWLPVEPKS CDKTHTCPPC PAPELLGGPS VFLFPPKPKD TLMISRTPEV TCVVVDVSHE DPEVKFNWYV DGVEVHNAKT KPREEQYNST YRVVSVLTVL HQDWLNGKEY KCKVSNKALP APIEKTISKA KGQPREPQVY TLPPSRDELT KNQVSLTCLV KGFYPSDIAV EWESNGQPEN NYKTTPPVLD SDGSFFLYSK LTVDKSRWQQ GNVFSCSVMH EALHNHYTQK SLSLSPGKHH HHHH | |
Greater than 85.0% as determined by SDS-PAGE. |