missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL17RB Protein

A cDNA sequence encoding the IL17RB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12409-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990329

Quantity Price
1 114.77 € / 2µg
10 107.33 € / 2µg
Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

55540
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
IL17RB
Human
His
IL17RB protein solution (0.25 mg/ml) containing phosphate buffered saline (pH 7.4) and 10% glycerol.
IL17RB Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Insect
REPTVQCGSE TGPSPEWMLQ HDLIPGDLRD LRVEPVTTSV ATGDYSILMN VSWVLRADAS IRLLKATKIC VTGKSNFQSY SCVRCNYTEA FQTQTRPSGG KWTFSYIGFP VELNTVYFIG AHNIPNANMN EDGPSMSVNF TSPGCLDHIM KYKKKCVKAG SLWDPNITAC KKNEETVEVN FTTTPLGNRY MALIQHSTII GFSQVFEPHQ KKQTRASVVI PVTGDSEGAT VQLTPYFPTC GSDCIRHKGT VVLCPQTGVP FPLDNNKSKP GGWLPVEPKS CDKTHTCPPC PAPELLGGPS VFLFPPKPKD TLMISRTPEV TCVVVDVSHE DPEVKFNWYV DGVEVHNAKT KPREEQYNST YRVVSVLTVL HQDWLNGKEY KCKVSNKALP APIEKTISKA KGQPREPQVY TLPPSRDELT KNQVSLTCLV KGFYPSDIAV EWESNGQPEN NYKTTPPVLD SDGSFFLYSK LTVDKSRWQQ GNVFSCSVMH EALHNHYTQK SLSLSPGKHH HHHH
Videos
SDS
Documents
Certificates