missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL1RL1 Protein

A cDNA sequence encoding the IL1RL1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12413-HIS-EC-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990339

Quantity Price
1 171.64 € / 10µg
10 161.27 € / 10µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

9173
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
IL1RL1
Human
His
IL1RL1 protein solution is supplied in 25mM Sodium Acetate pH 4.8 and 50% glycerol.
IL1RL1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGFWILAILT ILMYSTAAKF SKQSWGLENE ALIVRCPRQG KPSYTVDWYY SQTNKSIPTQ ERNRVFASGQ LLKFLPAAVA DSGIYTCIVR SPTFNRTGYA NVTIYKKQSD CNVPDYLMYS TVSGSEKNSK IYCPTIDLYN WTAPLEWFKN CQALQGSRYR AHKSFLVIDN VMTEDAGDYT CKFIHNENGA NYSVTATRSF TVKDEQGFSL FPVIGAPAQN EIKEVEIGKN ANLTCSACFG KGTQFLAAVL WQLNGTKITD FGEPRIQQEE GQNQSFSNGL ACLDMVLRIA DVKEEDLLLQ YDCLALNLHG LRRHTVRLSR KNPIDHHSIY CIIAVCSVFL MLINVLVIIL KMFWIEATLL WRDIAKPYKT RNDGKLYDAY VVYPRNYKSS TDGASRVEHF VHQILPDVLE NKCGYTLCIY GRDMLPGEDV VTAVETNIRK SRRHIFILTP QITHNKEFAY EQEVALHCAL IQNDAKVILI EMEALSELDM LQAEALQDSL QHLMKVQGTI KWREDHIANK RSLNSKFWKH VRYQMPVPSK IPRKASSLTP LAAQKQ
Videos
SDS
Documents
Certificates