missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL1RL1 Protein
A cDNA sequence encoding the IL1RL1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10726-10ug
Additional Details : Weight : 0.01000kg
Specifications
9173 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Measured by its ability to inhibit proliferation using D10. G4.1 mouse helper T cells. The ED50 for this effect is less or equal to 10ng/ml with IL-33. | |
Human | |
His | |
IL1RL1 protein solution (0.5 mg/ml) contains 10% glycerol and Phosphate Buffered Saline (pH 7.4). |
IL1RL1 Protein | |
Research Use Only | |
IL1RL1 | |
Recombinant Protein | |
Baculovirus | |
KFSKQSWGLE NEALIVRCPR QGKPSYTVDW YYSQTNKSIP TQERNRVFAS GQLLKFLPAA VADSGIYTCI VRSPTFNRTG YANVTIYKKQ SDCNVPDYLM YSTVSGSEKN SKIYCPTIDL YNWTAPLEWF KNCQALQGSR YRAHKSFLVI DNVMTEDAGD YTCKFIHNEN GANYSVTATR SFTVKDEQGF SLFPVIGAPA QNEIKEVEIG KNANLTCSAC FGKGTQFLAA VLWQLNGTKI TDFGEPRIQQ EEGQNQSFSN GLACLDMVLR IADVKEEDLL LQYDCLALNL HGLRRHTVRL SRKNPIDHHS LEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |