missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL34 Protein
A cDNA sequence encoding the IL34 was constructed and used to recombinantly synthesize the protein.
131.00 € - 6790.00 €
Specifications
Gene ID (Entrez) | 146433 |
---|---|
Name | IL34 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | IL34 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960369
|
enQuireBio™
QP12421-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940369
|
enQuireBio™
QP12421-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950369
|
enQuireBio™
QP12421-1MG |
1 mg |
6790.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
146433 | |
Research Use Only | |
IL34 | |
Human | |
His | |
The IL34 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
IL34 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMNE PLEMWPLTQN EECTVTGFLR DKLQYRSRLQ YMKHYFPINY KISVPYEGVF RIANVTRLQR AQVSERELRY LWVLVSLSAT ESVQDVLLEG HPSWKYLQEV QTLLLNVQQG LTDVEVSPKV ESVLSLLNAP GPNLKLVRPK ALLDNCFRVM ELLYCSCCKQ SSVLNWQDCE VPSPQSCSPE PSLQYAATQL YPPPPWSPSS PPHSTGSVRP VRAQGEGLLP | |
Greater than 85% as determined by SDS-PAGE. |