missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Immunoglobulin iota chain Protein

A DNA sequence encoding the Homo sapiens (Human) Immunoglobulin iota chain, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6882-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036530

  • 518.00 € / 100µg
Estimated Shipment: 28-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_009059.1
Immunoglobulin iota chain Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
7441
Greater than 90% as determined by SDS-PAGE.
Research Use Only
VPREB1
Recombinant Protein
E. coli
QPVLHQPPAM SSALGTTIRL TCTLRNDHDI GVYSVYWYQQ RPGHPPRFLL RYFSQSDKSQ GPQVPPRFSG SKDVARNRGY LSISELQPED EAMYYCAMGA RSSEKEERER EWEEEMEPTA ARTRVP
Videos
SDS
Documents
Certificates
Special Offers

Special Offers