missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Inactive phospholipase D5 Protein
A DNA sequence encoding the Homo sapiens (Human) Inactive phospholipase D5, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001182740.1 |
---|---|
Gene ID (Entrez) | 200150 |
Name | Inactive phospholipase D5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011361
|
enQuireBio™
QP7881-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031361
|
enQuireBio™
QP7881-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021361
|
enQuireBio™
QP7881-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041361
|
enQuireBio™
QP7881-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051361
|
enQuireBio™
QP7881-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061361
|
enQuireBio™
QP7881-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001182740.1 | |
Inactive phospholipase D5 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
200150 | |
Research Use Only | |
PLD5 | |
Recombinant Protein | |
E. coli | |
MGEDEDGLSE KNCQNKCRIA LVENIPEGLN YSENAPFHLS LFQGWMNLLN MAKKSVDIVS SHWDLNHTHP SACQGQRLFE KLLQLTSQNI EIKLVSDVTA DSKVLEALKL KGAEVTYMNM TAYNKGRLQS SFWIVDKQHV YIGSAGLDWQ SLGQMKELGV IFYNCSCLVL DLQRIFALYS SLKFKSRVPQ TWSKRLYGVY DNEKKLQLQL NETKSQAFVS NSPKLFCPKN RSFDIDAIYS VIDDAKQYVY IAVMDYLPIS STSTKRTYWP DLDAKIREAL VLRSVRVRLL LSFWKETDPL TFNFISSLKA ICTEIANCSL KVKFFDLERE NACATKEQKN HTFPRLNRNK YMVTDGAAYI GNFDWVGNDF TQNAGTGLVI NQADVRNNRS IIKQLKDVFE RDWYSPYAKT LQPTKQPNCS SLFKLKPLSN KTATDDTGGK DPRNV | |
Greater than 90% as determined by SDS-PAGE. |