missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interferon alpha 10 / IFNA10 Protein
A DNA sequence encoding the Homo sapiens (Human) GDF-15, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_002162.1 |
---|---|
Gene ID (Entrez) | 3446 |
Name | Interferon alpha 10 / IFNA10 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945639
|
enQuireBio™
QP5000-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985639
|
enQuireBio™
QP5000-YE-500UG |
500 μg |
1665.00 €
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955639
|
enQuireBio™
QP5000-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15975639
|
enQuireBio™
QP5000-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15995639
|
enQuireBio™
QP5000-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15965639
|
enQuireBio™
QP5000-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002162.1 | |
Interferon alpha 10 / IFNA10 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3446 | |
Research Use Only | |
IFNA10 | |
Recombinant Protein | |
Yeast | |
CDLPQTHSLG NRRALILLGQ MGRISPFSCL KDRHDFRIPQ EEFDGNQFQK AQAISVLHEM IQQTFNLFST EDSSAAWEQS LLEKFSTELY QQLNDLEACV IQEVGVEETP LMNEDSILAV RKYFQRITLY LIERKYSPCA WEVVRAEIMR SLSFSTNLQK RLRRKD | |
Greater than 90% as determined by SDS-PAGE. |