missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interferon alpha-inducible protein 1
A DNA sequence encoding the Homo sapiens (Human) Interferon alpha-inducible protein 6, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_002029.3 |
---|---|
Gene ID (Entrez) | 2537 |
Name | Interferon alpha-inducible protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16097821
|
enQuireBio™
QP9195-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037831
|
enQuireBio™
QP9195-YE-500UG |
500 μg |
1665.00 €
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007831
|
enQuireBio™
QP9195-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027831
|
enQuireBio™
QP9195-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047831
|
enQuireBio™
QP9195-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017831
|
enQuireBio™
QP9195-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002029.3 | |
Interferon alpha-inducible protein 1 | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2537 | |
Research Use Only | |
IFI6 | |
Recombinant Protein | |
Yeast | |
GKKKCSESSD SGSGFWKALT FMAVGGGLAV AGLPALGFTG AGIAANSVAA SLMSWSAILN GGGVPAGGLV ATLQSLGAGG SSVVIGNIGA LMGYATHKYL DSEEDEE | |
Greater than 90% as determined by SDS-PAGE. |