missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IRF 5 Protein
A cDNA sequence encoding the IRF 5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12459-100ug
Additional Details : Weight : 0.01000kg
Specifications
3663 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSPPTL QPPTLQPPVV LGPPAPDPSP LAPPPGNPAG FRELLSEVLE PGPLPASLPP AGEQLLPDLL I | |
Greater than 85% as determined by SDS-PAGE. |
IRF 5 Protein | |
Research Use Only | |
IRF5 | |
Human | |
His | |
IRF5 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT. |