missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ITLN1 / Omentin Protein
Recombinant Protein
Brand: enQuireBio™ QP5262-2ug
Additional Details : Weight : 0.01000kg
Specifications
NP_060095.2 | |
ITLN1 / Omentin Protein | |
Research Use Only | |
ITLN1 | |
Human | |
Untagged | |
10 mM sodium phosphate and 5:1 mannitol to protein, pH 7.5 |
55600 | |
2 μg | |
Less than or equal to 5.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MNQLSFLLFL IATTRGWSTD EANTYFKEWT CSSSPSLPRS CKEIKDECPS AFDGLYFLRT ENGVIYQTFC DMTSGGGGWT LVASVHENDM RGKCTVGDRW SSQQGSKADY PEGDGNWANY NTFGSAEAAT SDDYKNPGYY DIQAKDLGIW HVPNKSPMQH WRNSSLLRYR TDTGFLQTLG HNLFGIYQKY PVKYGEGKCW TDNGPVIPVV YDFGDAQKTA SYYSPYGQRE FTAGFVQFRV FNNERAANAL CAGMRVTGCN TEHHCIGGGG YFPEASPQQC GDFSGFDWSG YGTHVGYSSS REITEAAVLL FYR | |
Greater than 90 as determined by SDS-PAGE. |