missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IZUMO4 / C19orf36 Protein
A DNA sequence encoding the Homo sapiens (Human) IZUMO4 / C19orf36, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001026905.2 |
---|---|
Gene ID (Entrez) | 113177 |
Name | IZUMO4 / C19orf36 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039910
|
enQuireBio™
QP7632-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059910
|
enQuireBio™
QP7632-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049910
|
enQuireBio™
QP7632-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069910
|
enQuireBio™
QP7632-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079910
|
enQuireBio™
QP7632-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089910
|
enQuireBio™
QP7632-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001026905.2 | |
IZUMO4 / C19orf36 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
113177 | |
Research Use Only | |
IZUMO4 | |
Recombinant Protein | |
E. coli | |
HGCLHCHSNF SKKFSFYRHH VNFKSWWVGD IPVSGALLTD WSDDTMKELH LAIPAKITRE KLDQVATAVY QMMDQLYQGK MYFPGYFPNE LRNIFREQVH LIQNAIIESR IDCQHRCGIF QYETISCNNC TDSHVACFGY NCESSAQWKS AVQGLLNYIN NWHKQDTSMS LVSPALRCLE PPHLANLTLE DAAECLKQH | |
Greater than 90% as determined by SDS-PAGE. |