missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human JAZF1 Protein
A cDNA sequence encoding the JAZF1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12469-5ug
Additional Details : Weight : 0.01000kg
Specifications
JAZF1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTGIAAA SFFSNTCRFG GCGLHFPTLA DLIEHIEDNH IDTDPRVLEK QELQQPTYVA LSYINRFMTD AARREQESLK KKIQPKLSLT LSSSVSRGNV STPPRHSSGS LTPPVTPPIT PSSSFRSSTP TGSEYDEEEV DYEESDSDES WTTESAISSE AILSSMCMNG GEEKPFACPV PGCKKRYKNV NGIKYHAKNG HRTQIRVRKP FKCRCGKSYK TAQGLRHHTI NFHPPVSAEI IRKMQQ | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
JAZF1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |