missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Keratin, type I cuticular Ha5 Protein
A DNA sequence encoding the Homo sapiens (Human) Keratin, type I cuticular Ha5, was expressed in the hosts and tags indicated.
287.00 € - 3175.00 €
Specifications
Accession Number | NP_002271.3 |
---|---|
Gene ID (Entrez) | 3886 |
Name | Keratin, type I cuticular Ha5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16092061
|
enQuireBio™
QP7995-YE-100UG |
100 μg |
880.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032071
|
enQuireBio™
QP7995-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16002071
|
enQuireBio™
QP7995-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16022071
|
enQuireBio™
QP7995-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16042071
|
enQuireBio™
QP7995-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012071
|
enQuireBio™
QP7995-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002271.3 | |
Keratin, type I cuticular Ha5 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3886 | |
Research Use Only | |
KRT35 | |
Recombinant Protein | |
Yeast | |
MASKCLKAGF SSGSLKSPGG ASGGSTRVSA MYSSSSCKLP SLSPVARSFS ACSVGLGRSS YRATSCLPAL CLPAGGFATS YSGGGGWFGE GILTGNEKET MQSLNDRLAG YLEKVRQLEQ ENASLESRIR EWCEQQVPYM CPDYQSYFRT IEELQKKTLC SKAENARLVV EIDNAKLAAD DFRTKYETEV SLRQLVESDI NGLRRILDDL TLCKSDLEAQ VESLKEELLC LKKNHEEEVN SLRCQLGDRL NVEVDAAPPV DLNRVLEEMR CQYETLVENN RRDAEDWLDT QSEELNQQVV SSSEQLQSCQ AEIIELRRTV NALEIELQAQ HSMRDALEST LAETEARYSS QLAQMQCMIT NVEAQLAEIR ADLERQNQEY QVLLDVRARL ECEINTYRGL LESEDSKLPC NPCAPDYSPS KSCLPCLPAA SCGPSAARTN CSPRPICVPC PGGRF | |
Greater than 90% as determined by SDS-PAGE. |