missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Kita-kyushu lung cancer antigen 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Kita-kyushu lung cancer antigen 1, was expressed in the hosts and tags indicated.
202.00 € - 2720.00 €
Specifications
Accession Number | NP_001017978.1 |
---|---|
Gene ID (Entrez) | 203413 |
Name | Kita-kyushu lung cancer antigen 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16030151
|
enQuireBio™
QP7682-EC-100UG |
100 μg |
630.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050151
|
enQuireBio™
QP7682-EC-1MG |
1 mg |
2720.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040151
|
enQuireBio™
QP7682-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060151
|
enQuireBio™
QP7682-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070151
|
enQuireBio™
QP7682-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080151
|
enQuireBio™
QP7682-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001017978.1 | |
Kita-kyushu lung cancer antigen 1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-B2M | |
Tris-based buffer, 50% glycerol |
203413 | |
Research Use Only | |
CT83 | |
Recombinant Protein | |
E. coli | |
MNFYLLLASS ILCALIVFWK YRRFQRNTGE MSSNSTALAL VRPSSSGLIN SNTDNNLAVY DLSRDILNNF PHSIARQKRI LVNLSMVENK LVELEHTLLS KGFRGASPHR KST | |
Greater than 80% as determined by SDS-PAGE. |