missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLK11 Protein
A cDNA sequence encoding the KLK11 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12502-20ug
Additional Details : Weight : 0.01000kg
Specifications
11012 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MIIKGFECKP HSQPWQAALF EKTRLLCGAT LIAPRWLLTA AHCLKPRYIV HLGQHNLQKE EGCEQTRTAT ESFPHPGFNN SLPNKDHRND IMLVKMASPV SITWAVRPLT LSSRCVTAGT SCLISGWGST SSPQLRLPHT LRCANITIIE HQKCENAYPG NITDTMVCAS VQEGGKDSC GDSGGPLVCN QSLQGIISWG QDPCAITRKP GVYTKVCKYV DWIQET | |
Greater than 85.0% as determined by SDS-PAGE. |
KLK11 Protein | |
Research Use Only | |
KLK11 | |
Human | |
Untagged | |
KLK11 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |