missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLK3 Protein
A cDNA sequence encoding the KLK3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12506-HIS-1mg
Additional Details : Weight : 0.01000kg
Specifications
354 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
IVGGWECEKH SQPWQVLVAS RGRAVCGGVL VHPQWVLTAA HCIRNKSVIL LGRHSLFHPE DTGQVFQVSH SFPHPLYDMS LLKNRFLRPG DDSSHDLMLL RLSEPAELTD AVKVMDLPTQ EPALGTTCYA SGWGSIEPEE FLTPKKLQCV DLHVISNDVC AQVHPQKVTK FMLCAGRWTG GKSTCSGDSG GPLVCNGVLQ GITSWGSEPC ALPERPSLYT KVVHYRKWIK DTIVANP | |
Greater than 90% as determined by SDS-PAGE. |
KLK3 Protein | |
Research Use Only | |
KLK3 | |
Human | |
His | |
KLK3 protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl pH 7.4. |