missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LAIR1 Protein

A cDNA sequence encoding the LAIR1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12526-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990709

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

LAIR1 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The protein (1 mg/ml) contains 20mM Tris-HCl (pH 7.5) 50mM NaCl and 1mM EDTA.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MQEEDLPRPS ISAEPGTVIP LGSHVTFVCR GPVGVQTFRL ERESRSTYND TEDVSQASPSESEARFRIDS VSEGNAGPYR CIYYKPPKWS EQSDYLELLV KETSG
Videos
SDS
Documents
Certificates