missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LAMP1 Protein
A cDNA sequence encoding the LAMP1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | LAMP1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15980719
|
enQuireBio™
QP12529-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960719
|
enQuireBio™
QP12529-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970719
|
enQuireBio™
QP12529-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
LAMP1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
LAMP1 protein solution (1 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPAMFMVKN GNGTACIMAN FSAAFSVNYD TKSGPKNMTF DLPSDATVVL NRSSCGKENT SDPSLVIAFG RGHTLTLNFT RNATRYSVQL MSFVYNLSDT HLFPNASSKE IKTVESITDI RADIDKKYRC VSGTQVHMNN VTVTLHDATI QAYLSNSSFS RGETRCEQDR PSPTTAPPAPPSPSPSPVPK SPSVDKYNVS GTNGTCLLAS MGLQLNLTYE RKDNTTVTRL LNINPNKTSA SGSCGAHLVT LELHSEGTTV LLFQFGMNAS SSRFFLQGIQ LNTILPDARD PAFKAANGSL RALQATVGNS YKCNAEEHVR VTKAFSVNIF KVWVQAFKVE GGQFGSVEEC LLDENSMHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |