missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LBH Protein
A cDNA sequence encoding the LBH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12543-10ug
Additional Details : Weight : 0.01000kg
Specifications
81606 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSIYFPI HCPDYLRSAK MTEVMMNTQP MEEIGLSPRK DGLSYQIFPD PSDFDRCCKL KDRLPSIVVE PTEGEVESGE LRWPPEEFLV QEDEQDNCEE TAKENKEQ | |
Greater than 90.0% as determined by SDS-PAGE. |
LBH Protein | |
Research Use Only | |
LBH | |
Human | |
His | |
LBH protein solution (0.25 mg/ml) containing 20mM Tris-HCl(pH 8.0), 20% glycerol, 0.15M NaCl and 1mM DTT. |