missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LCAT Protein

A DNA sequence encoding the Homo sapiens (Human) LCAT, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8712-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046081

  • 518.00 € / 100µg
Estimated Shipment: 05-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000220.1
LCAT Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
3931
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LCAT
Recombinant Protein
E. coli
FWLLNVLFPP HTTPKAELSN HTRPVILVPG CLGNQLEAKL DKPDVVNWMC YRKTEDFFTI WLDLNMFLPL GVDCWIDNTR VVYNRSSGLV SNAPGVQIRV PGFGKTYSVE YLDSSKLAGY LHTLVQNLVN NGYVRDETVR AAPYDWRLEP GQQEEYYRKL AGLVEEMHAA YGKPVFLIGH SLGCLHLLYF LLRQPQAWKD RFIDGFISLG APWGGSIKPM LVLASGDNQG IPIMSSIKLK EEQRITTTSP WMFPSRMAWP EDHVFISTPS FNYTGRDFQR FFADLHFEEG WYMWLQSRDL LAGLPAPGVE VYCLYGVGLP TPRTYIYDHG FPYTDPVGVL YEDGDDTVAT RSTELCGLWQ GRQPQPVHLL PLHGIQHLNM VFSNLTLEHI NAILLGAYRQ GPPASPTAS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers