missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LCN1 Protein
A cDNA sequence encoding the LCN1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 3933 |
---|---|
Name | LCN1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | LCN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960769
|
enQuireBio™
QP12545-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950769
|
enQuireBio™
QP12545-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940769
|
enQuireBio™
QP12545-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
3933 | |
Research Use Only | |
LCN1 | |
Human | |
His | |
LCN1 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |
LCN1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMHHLLA SDEEIQDVSG TWYLKAMTVD REFPEMNLES VTPMTLTTLE GGNLEAKVTM LISGRCQEVK AVLEKTDEPG KYTADGGKHV AYIIRSHVKD HYIFYCEGEL HGKPVRGVKL VGRDPKNNLE ALEDFEKAAG ARGLSTESIL IPRQSETCSP GSD | |
Greater than 90.0% as determined by SDS-PAGE. |