missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LELP1 Protein
A DNA sequence encoding the Homo sapiens (Human) LELP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7697-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001010857.1 | |
LELP1 Protein | |
Research Use Only | |
LELP1 | |
Recombinant Protein | |
E. coli | |
MSSDDKSKSN DPKTEPKNCD PKCEQKCESK CQPSCLKKLL QRCFEKCPWE KCPAPPKCLP CPSQSPSSCP PQPCTKPCPP KCPSSCPHAC PPPCPPPE | |
Greater than 80% as determined by SDS-PAGE. |
149018 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |