missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Leptin Protein
A DNA sequence encoding the Human Leptin Protein, was expressed in the hosts and tags indicated.
105.00 € - 373.00 €
Specifications
Accession Number | NP_000221.1 |
---|---|
Gene ID (Entrez) | 3952 |
Name | Leptin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than 1.0 EU/ug as determined by LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15989078
|
enQuireBio™
QP1201-EC-1MG |
1 mg |
373.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999078
|
enQuireBio™
QP1201-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000221.1 | |
Leptin Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
VPIQKVQDDT KTLIKTIVTR INDISHTQSV SSKQKVTGLD FIPGLHPILT LSKMDQTLAV YQQILTSMPS RNVIQISNDL ENLRDLLHVL AFSKSCHLPW ASGLETLDSL GGVLEASGYS TEVVALSRLQ GSLQDMLWQL DLSPGC | |
Greater than 97% as determined by SDS-PAGE and HPLC. |
3952 | |
Research Use Only | |
LEP | |
The ED50 as determined by a chemotaxis bioassay using human Leptin R transfected BaF3 murine proB cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |