missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Leptin Protein

A DNA sequence encoding the Human Leptin Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP1201-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989078

Quantity Price
1 318.47 € / 1mg
6 299.84 € / 1mg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NP_000221.1
Leptin Protein
1 mg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
VPIQKVQDDT KTLIKTIVTR INDISHTQSV SSKQKVTGLD FIPGLHPILT LSKMDQTLAV YQQILTSMPS RNVIQISNDL ENLRDLLHVL AFSKSCHLPW ASGLETLDSL GGVLEASGYS TEVVALSRLQ GSLQDMLWQL DLSPGC
3952
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
LEP
The ED50 as determined by a chemotaxis bioassay using human Leptin R transfected BaF3 murine proB cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
SDS
Documents
Certificates