missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Leuprorelin Protein
Recombinant Protein
131.00 € - 955.00 €
Specifications
Name | Leuprorelin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960799
|
enQuireBio™
QP12555-5MG |
5 mg |
131.00 €
5mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950799
|
enQuireBio™
QP12555-25MG |
25 mg |
336.00 €
25mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940799
|
enQuireBio™
QP12555-100MG |
100 mg |
955.00 €
100mg |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Leuprorelin Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The Leuprorelin was lyophilized from a concentrated (1 mg/ml) solution with no additives. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
pGlu-His-Trp-Ser-Tyr-D-Leu-Leu-Arg-Pro-NHEt | |
Greater than 98.0% as determined by RP-HPLC. |