missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LFA 3 Protein
A cDNA sequence encoding the LFA 3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12556-20ug
Additional Details : Weight : 0.01000kg
Specifications
965 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MVAGSDAGRA LGVLSVVCLL HCFGFISCFS QQIYGVVYGN VTFHVPSNVP LKEVLWKKQK DKVAELENSE FRAFSSFKNR VYLDTVSGSL TIYNLTSSDE DEYEMESPNI TDTMKFFLYV LESLPSPTLT CALTNGSIEV QCMIPEHYNS HRGLIMYSWD CPMEQCKRNS TSIYFKMEND LPQKIQCTLS NPLFNTTSSI ILTTCIPSSG HSRHRYALIP IPLAVITTCI VLYMNGILKC DRKPDRTNSN | |
Greater than 98.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
LFA 3 Protein | |
Research Use Only | |
CD58 | |
Human | |
Untagged | |
Each mg of CD58 contains 0.8 mg sucrose, 0.3 mg glycine, 0.25 mg sodium citrate dihydrate, and 4microg citric acid monohydrate. |