missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LFA 3 Protein
A cDNA sequence encoding the LFA 3 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1245.00 €
Specifications
Gene ID (Entrez) | 965 |
---|---|
Name | LFA 3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD58 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15990799
|
enQuireBio™
QP12556-20UG |
20 μg |
131.00 €
20µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970799
|
enQuireBio™
QP12556-100UG |
100 μg |
203.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15980799
|
enQuireBio™
QP12556-1MG |
1 mg |
1245.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
965 | |
Research Use Only | |
CD58 | |
Human | |
Untagged | |
Each mg of CD58 contains 0.8 mg sucrose, 0.3 mg glycine, 0.25 mg sodium citrate dihydrate, and 4microg citric acid monohydrate. |
LFA 3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MVAGSDAGRA LGVLSVVCLL HCFGFISCFS QQIYGVVYGN VTFHVPSNVP LKEVLWKKQK DKVAELENSE FRAFSSFKNR VYLDTVSGSL TIYNLTSSDE DEYEMESPNI TDTMKFFLYV LESLPSPTLT CALTNGSIEV QCMIPEHYNS HRGLIMYSWD CPMEQCKRNS TSIYFKMEND LPQKIQCTLS NPLFNTTSSI ILTTCIPSSG HSRHRYALIP IPLAVITTCI VLYMNGILKC DRKPDRTNSN | |
Greater than 98.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |