missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LFNG Protein
A DNA sequence encoding the Homo sapiens (Human) LFNG, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001035257.1 |
---|---|
Gene ID (Entrez) | 3955 |
Name | LFNG Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011511
|
enQuireBio™
QP7905-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031511
|
enQuireBio™
QP7905-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021511
|
enQuireBio™
QP7905-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041511
|
enQuireBio™
QP7905-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051511
|
enQuireBio™
QP7905-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061511
|
enQuireBio™
QP7905-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001035257.1 | |
LFNG Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3955 | |
Research Use Only | |
LFNG | |
Recombinant Protein | |
E. coli | |
MTPGRCCLAA DIQVETFIFT DGEDEALARH TGNVVITNCS AAHSRQALSC KMAVEYDRFI ESGRKWFCHV DDDNYVNLRA LLRLLASYPH TRDVYVGKPS LDRPIQAMER VSENKVRPVH FWFATGGAGF CISRGLALKM SPWASGGHFM NTAERIRLPD DCTIGYIVEA LLGVPLIRSG LFHSHLENLQ QVPTSELHEQ VTLSYGMFEN KRNAVHVKGP FSVEADPSRF RSIHCHLYPD TPWCPRTAIF | |
Greater than 80% as determined by SDS-PAGE. |