missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LGALS14 Protein

A cDNA sequence encoding the LGALS14 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12560-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990809

Quantity Price
1 114.77 € / 1µg
10 107.33 € / 1µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

LGALS14 Protein
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The LGALS14 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSSLPVP YTLPVSLPVG SCVIITGTPI LTFVKDPQLE VNFYTGMDED SDIAFQFRLH FGHPAIMNSR VFGIWRYEEK CYYLPFEDGK PFELCIYVRH KEYKVMVNGQ RIYNFAHRFP PASVKMLQVL RDISLTRVLI SD
Videos
SDS
Documents
Certificates