missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LHRH Protein
A cDNA sequence encoding the LHRH was constructed and used to recombinantly synthesize the protein.
131.00 € - 481.00 €
Specifications
Gene ID (Entrez) | 2796 |
---|---|
Name | LHRH Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | GNRH1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15970839
|
enQuireBio™
QP12569-10MG |
10 mg |
131.00 €
10mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15980839
|
enQuireBio™
QP12569-25MG |
25 mg |
179.00 €
25mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960839
|
enQuireBio™
QP12569-100MG |
100 mg |
481.00 €
100mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
2796 | |
Research Use Only | |
GNRH1 | |
Human | |
Untagged | |
The LHRH was lyophilized from a concentrated (1 mg/ml) solution with no additives. |
LHRH Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2 | |
Greater than 98.0% as determined by RP-HPLC. |