missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LIM domain transcription factor LMO4 Protein
A DNA sequence encoding the Homo sapiens (Human) LIM domain transcription factor LMO4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6313-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006760.1 | |
LIM domain transcription factor LMO4 Protein | |
Research Use Only | |
LMO4 | |
Recombinant Protein | |
E. coli | |
MVNPGSSSQP PPVTAGSLSW KRCAGCGGKI ADRFLLYAMD SYWHSRCLKC SCCQAQLGDI GTSCYTKSGM ILCRNDYIRL FGNSGACSAC GQSIPASELV MRAQGNVYHL KCFTCSTCRN RLVPGDRFHY INGSLFCEHD RPTALINGHL NSLQSNPLLP DQKVC | |
Greater than 80% as determined by SDS-PAGE. |
8543 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |