missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LIM and SH3 domain protein 1
A DNA sequence encoding the Homo sapiens (Human) LIM and SH3 domain protein 1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001258537.1 |
---|---|
Gene ID (Entrez) | 3927 |
Name | LIM and SH3 domain protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044841
|
enQuireBio™
QP8479-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064841
|
enQuireBio™
QP8479-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054841
|
enQuireBio™
QP8479-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074841
|
enQuireBio™
QP8479-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084841
|
enQuireBio™
QP8479-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094841
|
enQuireBio™
QP8479-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001258537.1 | |
LIM and SH3 domain protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3927 | |
Research Use Only | |
LASP1 | |
Recombinant Protein | |
E. coli | |
MNPNCARCGK IVYPTEKVNC LDKFWHKACF HCETCKMTLN MKNYKGYEKK PYCNAHYPKQ SFTMVADTPE NLRLKQQSEL QSQVRYKEEF EKNKGKGFSV VADTPELQRI KKTQDQISNI KYHEEFEKSR MGPSGGEGME PERRDSQDGS SYRRPLEQQQ PHHIPTSAPV YQQPQQQPVA QSYGGYKEPA APVSIQRSAP GGGGKRYRAV YDYSAADEDE VSFQDGDTIV NVQQIDDGWM YGT | |
Greater than 90% as determined by SDS-PAGE. |