missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LIN7A Protein
A cDNA sequence encoding the LIN7A was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | LIN7A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15950859
|
enQuireBio™
QP12573-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930859
|
enQuireBio™
QP12573-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940859
|
enQuireBio™
QP12573-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
LIN7A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
LIN7A protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLKPSVT SAPTADMATL TVVQPLTLDR DVARAIELLE KLQESGEVPV HKLQSLKKVL QSEFCTAIRE VYQYMHETIT VNGCPEFRAR ATAKATVAAF AASEGHSHPR VVELPKTDEG LGFNVMGGKE QNSPIYISRI IPGGVAERHG GLKRGDQLLS VNGVSVEGEH HEKAVELLKA AKDSVKLVVR YTPKVLEEME ARFEKLRTAR RRQQQQLLIQ QQQQQQQQQT QQNHMS | |
Greater than 90.0% as determined by SDS-PAGE |