missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lipoyl synthase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Lipoyl synthase, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6307-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001265519.1 | |
Lipoyl synthase, mitochondrial Protein | |
Research Use Only | |
LIAS | |
Recombinant Protein | |
E. coli | |
LSSLPDKKKE LLQNGPDLQD FVSGDLADRS TWDEYKGNLK RQKGERLRLP PWLKTEIPMG KNYNKLKNTL RNLNLHTVCE EARCPNIGEC WGGGEYATAT ATIMLMGDTC TRGCRFCSVK TARNPPPLDA SEPYNTAKAI AEWGLDYVVL TSVDRDDMPD GGAEHIAKTV SYLKERNPKI LVECLTPDFR GDLKAIEKVA LSGLDVYAHN VETVPELQSK VRDPRANFDQ SLRVLKHAKK VQPDVISKTS IMLGLGENDE QVYATMKALR EADVDCLTLG QYMQPTRRHL KVEEYITPEK FKYWEKVGNE LGFHYTASGP LVRSSYKAGE FFLKNLVAKR KTKDL | |
Greater than 90% as determined by SDS-PAGE. |
11019 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |