missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LRPAP1 Protein
A cDNA sequence encoding the LRPAP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12585-20ug
Additional Details : Weight : 0.01000kg
Specifications
LRPAP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMYSREK NQPKPSPKRE SGEEFRMEKL NQLWEKAQRL HLPPVRLAEL HADLKIQERD ELAWKKLKLD GLDEDGEKEA RLIRNLNVIL AKYGLDGKKD ARQVTSNSLS GTQEDGLDDP RLEKLWHKAK TSGKFSGEEL DKLWREFLHH KEKVHEYNVL LETLSRTEEI HENVISPSDL SDIKGSVLHS RHTELKEKLR SINQGLDRLR RVSHQGYSTE AEFEEPRVID LWDLAQSANL TDKELEAFRE ELKHFEAKIE KHNHYQKQLE IAHEKLRHAE SVGDGERVSR SREKHALLEG RTKELGYTVK KHLQDLSGRI SRARHNEL | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The LRPAP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 20% glycerol. |