missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lutropin-choriogonadotropic hormone receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Lutropin-choriogonadotropic hormone receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6305-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000224.2 | |
Lutropin-choriogonadotropic hormone receptor Protein | |
Research Use Only | |
Lhcgr | |
Recombinant Protein | |
E. coli | |
EALCPEPCNC VPDGALRCPG PTAGLTRLSL AYLPVKVIPS QAFRGLNEVI KIEISQIDSL ERIEANAFDN LLNLSEILIQ NTKNLRYIEP GAFINLPRLK YLSICNTGIR KFPDVTKVFS SESNFILEIC DNLHITTIPG NAFQGMNNES VTLKLYGNGF EEVQSHAFNG TTLTSLELKE NVHLEKMHNG AFRGATGPKT LDISSTKLQA LPSYGLESIQ RLIATSSYSL KKLPSRETFV NLLEATLTYP SHCCAFRNLP TKEQNFSHSI SENFSKQCES TVRKVNNKTL YSSMLAESEL SGWDYEYGFC LPKTPRCAPE PDAFNPCEDI MGYDFLR | |
Greater than 90% as determined by SDS-PAGE. |
3973 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |