missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lysine-specific demethylase 5A Protein
A DNA sequence encoding the Homo sapiens (Human) Lysine-specific demethylase 5A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6257-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001036068.1 | |
Lysine-specific demethylase 5A Protein | |
Research Use Only | |
KDM5A | |
Recombinant Protein | |
Yeast | |
EYALSGWNLN NMPVLEQSVL AHINVDISGM KVPWLYVGMC FSSFCWHIED HWSYSINYLH WGEPKTWYGV PSHAAEQLEE VMRELAPELF ESQPDLLHQL VTIMNPNVLM EHGVPVYRTN QCAGEFVVTF PRAYHSGFNQ GYNFAEAVNF CTADWLPIGR QCVNHYR | |
Greater than 90% as determined by SDS-PAGE. |
5927 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |