missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human M2 Protein
A cDNA sequence encoding the M2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12615-5ug
Additional Details : Weight : 0.01000kg
Specifications
M2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
M2 is supplied in 16mM HEPES buffer pH 8.0, 400mM NaCl, and 20% glycerol. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Greater than 75% as determined by SDS-PAGE. |