missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human m7GpppN-mRNA hydrolase Protein
A DNA sequence encoding the Homo sapiens (Human) m7GpppN-mRNA hydrolase, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_001229306.1 |
---|---|
Gene ID (Entrez) | 167227 |
Name | m7GpppN-mRNA hydrolase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16050931
|
enQuireBio™
QP7809-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090931
|
enQuireBio™
QP7809-YE-500UG |
500 μg |
1665.00 €
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060931
|
enQuireBio™
QP7809-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080931
|
enQuireBio™
QP7809-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000941
|
enQuireBio™
QP7809-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070931
|
enQuireBio™
QP7809-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001229306.1 | |
m7GpppN-mRNA hydrolase Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
167227 | |
Research Use Only | |
DCP2 | |
Recombinant Protein | |
Yeast | |
METKRVEIPG SVLDDFCSRF ILHIPSEERD NAIRVCFQIE LAHWFYLDFY MQNTPGLPQC GIRDFAKAVF SHCPFLLPQG EDVEKVLDEW KEYKMGVPTY GAIILDETLE NVLLVQGYLA KSGWGFPKGK VNKEEAPHDC AAREVFEETG FDIKDYICKD DYIELRINDQ LARLYIIPGI PKDTKFNPKT RREIRNIEWF SIEKLPCHRN DMTPKSKLGL APNKFFMAIP FIRPLRDWLS RRFGDSSDSD NGFSSTGSTP AKPTVEKLSR TKFRHSQQLF PDGSPGDQWV KHRQPLQQKP YNNHSEMSDL LKGKKCEKKL HPRKLQDNFE TDAVYDLPSS SEDQLLEHAE GQPVACNGHC KFPFSSRAFL SFKFDHNAIM KILDL | |
Greater than 90% as determined by SDS-PAGE. |