missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAFF Protein
A cDNA sequence encoding the MAFF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12619-1mg
Additional Details : Weight : 0.01000kg
Specifications
MAFF Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSVDPLS SKALKIKREL SENTPHLSDE ALMGLSVREL NRHLRGLSAE EVTRLKQRRR TLKNRGYAAS CRVKRVCQKE ELQKQKSELE REVDKLAREN AAMRLELDAL RGKCEALQGF ARSVAAARGP ATLVAPASVI TIVKSTPGSG SGPAHGPDPA HGPASCS | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MAFF solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |