missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAPKAP1 Protein
A DNA sequence encoding the Homo sapiens (Human) MAPKAP1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001006618.1 |
---|---|
Gene ID (Entrez) | 79109 |
Name | MAPKAP1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16042141
|
enQuireBio™
QP8011-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16062141
|
enQuireBio™
QP8011-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16052141
|
enQuireBio™
QP8011-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072141
|
enQuireBio™
QP8011-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16082141
|
enQuireBio™
QP8011-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092141
|
enQuireBio™
QP8011-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001006618.1 | |
MAPKAP1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
79109 | |
Research Use Only | |
MAPKAP1 | |
Recombinant Protein | |
E. coli | |
AFLDNPTIIL AHIRQSHVTS DDTGMCEMVL IDHDVDLEKI HPPSMPGDSG SEIQGSNGET QGYVYAQSVD ITSSWDFGIR RRSNTAQRLE RLRKERQNQI KCKNIQWKER NSKQSAQELK SLFEKKSLKE KPPISGKQSI LSVRLEQCPL QLNNPFNEYS KFDGKGHVGT TATKKIDVYL PLHSSQDRLL PMTVVTMASA RVQDLIGLIC WQYTSEGREP KLNDNVSAYC LHIAEDDGEV DTDFPPLDSN EPIHKFGFST LALVEKYSSP GLTSKESLFV RINAAHGFSL IQVDNTKVTM KEILLKAVKR RKGSQKVSGP QYRLEKQSEP NVAVDLDSTL ESQSAWEFCL VRENSSRADG VFEEDSQIDI ATVQDMLSSH HYKSFKVSMI HRLRFTTDVQ LGISGDKVEI DPVTNQKAST KFWIKQKPIS IDSDLLCACD LAEEKSPSHA IFKLTYLSNH DYKHLYFESD AATVNEIVLK VNYILESRAS TARADYFAQK QRKLNRRTSF SFQKEKKSGQ Q | |
Greater than 90% as determined by SDS-PAGE. |