missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MBD3 Protein

A cDNA sequence encoding the MBD3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12654-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991119

Quantity Price
1 114.77 € / 2µg
10 107.33 € / 2µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

MBD3 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The MBD3 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 1mM DTT, 1mM EDTA and 40% glycerol.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMERKRWE CPALPQGWER EEVPRRSGLS AGHRDVFYYS PSGKKFRSKP QLARYLGGSM DLSTFDFRTG KMLMSKMNKS RQRVRYDSSN QVKGKPDLNT ALPVRQTASI FKQPVTKITN HPSNKVKSDP QKAVDQPRQL FWEKKLSGLN AFDIAEELVK TMDLPKGLQG VGPGCTDETL LSAIASALHT STMPITGQLS AAVEKNPGVW LNTTQPLCKA FMVTDEDIRK QEELVQQVRK RLEEALMADM LAHVEELARD GEAPLDKACA EDDDEEDEEE EEEEPDPDPE MEHV
Videos
SDS
Documents
Certificates