missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MED4 Protein

A cDNA sequence encoding the MED4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12673-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991179

  • 203.00 € / 25µg
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MED4 Protein
Research Use Only
Recombinant Protein
E. coli
MAASSSGEKE KERLGGGLGV AGGNSTRERL LSALEDLEVL SRELIEMLAI SRNQKLLQAG EENQVLELLI HRDGEFQELM KLALNQGKIH HEMQVLEKEV EKRDGDIQQL QKQLKEAEQI LATAVYQAKE KLKSIEKARK GAISSEEIIK YAHRISASNA VCAPLTWVPG DPRRPYPTDL EMRSGLLGQM NNPSTNGVNG HLPGDALAAG RLPDVLAPQY PWQSNDMSMN MLPPNHSSDF LLEPPGHNKE DEDDVEIMST DSSSSSSESD LEHHHHHH
Greater than 80.0% as determined by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The MED4 protein solution contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 1mM DTT and 100mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers