missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Metaxin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Metaxin-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6393-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001006636.1 | |
Metaxin-2 Protein | |
Research Use Only | |
MTX2 | |
Recombinant Protein | |
E. coli | |
MSLVAEAFVS QIAAAEPWPE NATLYQQLKG EQILLSDNAA SLAVQAFLQM CNLPIKVVCR ANAEYMSPSG KVPFIHVGNQ VVSELGPIVQ FVKAKGHSLS DGLEEVQKAE MKAYMELVNN MLLTAELYLQ WCDEATVGEI THARYGSPYP WPLNHILAYQ KQWEVKRKMK AIGWGKKTLD QVLEDVDQCC QALSQRLGTQ PYFFNKQPTE LDALVFGHLY TILTTQLTND ELSEKVKNYS NLLAFCRRIE QHYFEDRGKG RLS | |
Greater than 80% as determined by SDS-PAGE. |
10651 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |