missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mixed lineage kinase domain-like Protein
A DNA sequence encoding the Homo sapiens (Human) Mixed lineage kinase domain-like protein, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7971-ec-10ug
Informations supplémentaires : Weight : 0.01000kg
Spécification
NP_001135969.1 | |
Mixed lineage kinase domain-like Protein | |
Research Use Only | |
MLKL | |
Recombinant Protein | |
E. coli | |
MENLKHIITL GQVIHKRCEE MKYCKKQCRR LGHRVLGLIK PLEMLQDQGK RSVPSEKLTT AMNRFKAALE EANGEIEKFS NRSNICRFLT ASQDKILFKD VNRKLSDVWK ELSLLLQVEQ RMPVSPISQG ASWAQEDQQD ADEDRRAFQM LRRDNEKIEA SLRRLEINMK EIKETLRQYL PPKCMQEIPQ EQIKEIKKEQ LSGSPWILLR ENEVSTLYKG EYHRAPVAIK VFKKLQAGSI AIVRQTFNKE IKTMKKFESP NILRIFGICI DETVTPPQFS IVMEYCELGT LRELLDREKD LTLGKRMVLV LGAARGLYRL HHSEAPELHG KIRSSNFLVT QGYQVKLAGF ELRKTQTSMS LGTTREKTDR VKSTAYLSPQ ELEDVFYQYD VKSEIYSFGI VLWEIATGDI PFQGCNSEKI RKLVAVKRQQ EPLGEDCPSE LREIIDECRA HDPSVRPSVD EILKKLSTFS K | |
Greater than 90% as determined by SDS-PAGE. |
197259 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |