missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MKK6 / MEK6 / MAP2K6 Protein
A DNA sequence encoding the Homo sapiens (Human) MKK6 / MEK6 / MAP2K6, was expressed in the hosts and tags indicated.
530.00 € - 1475.00 €
Specifications
Accession Number | NP_002749.2 |
---|---|
Gene ID (Entrez) | 5608 |
Name | MKK6 / MEK6 / MAP2K6 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16042580
|
enQuireBio™
QP6338-BA-20UG |
20 μg |
530.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032580
|
enQuireBio™
QP6338-BA-100UG |
100 μg |
1475.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16052580
|
enQuireBio™
QP6338-BA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002749.2 | |
MKK6 / MEK6 / MAP2K6 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5608 | |
Research Use Only | |
MAP2K6 | |
Recombinant Protein | |
Baculovirus | |
MSQSKGKKRN PGLKIPKEAF EQPQTSSTPP RDLDSKACIS IGNQNFEVKA DDLEPIMELG RGAYGVVEKM RHVPSGQIMA VKRIRATVNS QEQKRLLMDL DISMRTVDCP FTVTFYGALF REGDVWICME LMDTSLDKFY KQVIDKGQTI PEDILGKIAV SIVKALEHLH SKLSVIHRDV KPSNVLINAL GQVKMCDFGI SGYLVDSVAK TIDAGCKPYM APERINPELN QKGYSVKSDI WSLGITMIEL AILRFPYDSW GTPFQQLKQV VEEPSPQLPA DKFSAEFVDF TSQCLKKNSK ERPTYPELMQ HPFFTLHESK GTDVASFVKL ILGD | |
Greater than 90% as determined by SDS-PAGE. |