missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MLLT11 Protein
A cDNA sequence encoding the MLLT11 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12701-20ug
Additional Details : Weight : 0.01000kg
Specifications
MLLT11 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMRDPVSS QYSSFLFWRM PIPELDLSEL EGLGLSDTAT YKVKDSSVGK MIGQATAADQ EKNPEGDGLL EYSTFNFWRA PIASIHSFEL DLL | |
Greater than 80.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MLLT11 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |